Assay Details
| Gene Symbol: C5orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294337.1 | NP_001281266.1 | 1 - 2 | 341 | 128 | |
| GenBank mRNA | BX090247.1 | - | 1 - 2 | 129 | ||
More Information
Set Membership: |
|

C5orf38
Genomic Map