| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000305108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000305108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000305108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000305108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000305108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000305108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000305108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000305108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000305108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000305108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000397265.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000397265.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000397265.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000397265.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000397265.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000397265.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000397265.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000433832.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000433832.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000433832.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000433832.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000433832.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000433832.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000537416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000537416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000537416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000537416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000542920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000542920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000542920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000542920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000542920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000542920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
ENST00000542920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_001294346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_016533.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_016533.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_016533.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_016533.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_016533.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_016533.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_016533.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_016533.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009558.hg.1 |
NM_016533.5 |
Refseq |