Assay Details
| Gene Symbol: TTC39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080494.3 | NP_001073963.1 | 16 - 17 | 1603 | 113 | |
| NM_001144832.2 | NP_001138304.1 | 16 - 17 | 1578 | 113 | ||
| NM_001297663.1 | NP_001284592.1 | 16 - 17 | 1580 | 113 | ||
| NM_001297664.1 | NP_001284593.1 | 16 - 17 | 1616 | 113 | ||
| NM_001297665.1 | NP_001284594.1 | 16 - 17 | 1708 | 113 | ||
| NM_001297666.1 | NP_001284595.1 | 5 - 6 | 611 | 113 | ||
| NM_001297667.1 | NP_001284596.1 | 4 - 5 | 519 | 113 | ||
| XM_005270644.2 | XP_005270701.1 | 16 - 17 | 1609 | 113 | ||
| XM_006710471.2 | XP_006710534.1 | 16 - 17 | 1706 | 113 | ||
| XM_011541048.2 | XP_011539350.1 | 16 - 17 | 1683 | 113 | ||
| XM_011541049.2 | XP_011539351.1 | 16 - 17 | 1822 | 113 | ||
| XM_011541050.2 | XP_011539352.1 | 16 - 17 | 1683 | 113 | ||
| XM_011541051.2 | XP_011539353.1 | 16 - 17 | 1716 | 113 | ||
| XM_011541052.2 | XP_011539354.1 | 16 - 17 | 1662 | 113 | ||
| XM_011541053.2 | XP_011539355.1 | 16 - 17 | 1700 | 113 | ||
| XM_011541054.2 | XP_011539356.1 | 16 - 17 | 1536 | 113 | ||
| XM_017000715.1 | XP_016856204.1 | 16 - 17 | 1583 | 113 | ||
| GenBank mRNA | AF007170.1 | - | 16 - 17 | 1484 | 113 | |
| AK298019.1 | - | 16 - 17 | 1587 | 113 | ||
| AK301587.1 | - | 16 - 17 | 1576 | 113 | ||
| BC004399.1 | - | 5 - 6 | 572 | 113 | ||
| BC028374.2 | - | 16 - 17 | 1650 | 113 | ||
| BQ227134.1 | - | 4 - 5 | 530 | 113 | ||
More Information
Set Membership: |
|

TTC39A
Genomic Map