Assay Details
| Gene Symbol: ART5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297668.1 | NP_001284597.1 | 2 - 3 | 1039 | 66 | |
| GenBank mRNA | AY358466.1 | - | 2 - 3 | 664 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

ART5
Genomic Map