| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009017.hg.1 |
ENST00000605147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009017.hg.1 |
ENST00000605147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009017.hg.1 |
NR_104273.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009017.hg.1 |
NR_104273.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009017.hg.1 |
ENST00000605147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009017.hg.1 |
ENST00000605147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009017.hg.1 |
NR_104273.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009017.hg.1 |
NR_104273.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161835.hg.1 |
ENST00000605147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161835.hg.1 |
NR_104273.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161835.hg.1 |
ENST00000605147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161835.hg.1 |
NR_104273.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161835.hg.1 |
TC0400009017.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400161836.hg.1 |
ENST00000605147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161836.hg.1 |
NR_104273.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161836.hg.1 |
ENST00000605147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161836.hg.1 |
NR_104273.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161836.hg.1 |
TC0400009017.hg.1 |
Clariom D TC |