| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
ENST00000622141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009039.hg.1 |
NR_106864.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900171437.hg.1 |
ENST00000622141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900171437.hg.1 |
NR_106864.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900171437.hg.1 |
TC1900009039.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900171437.hg.1 |
ENST00000622141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900171437.hg.1 |
NR_106864.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
ENST00000329665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
ENST00000622141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
NM_181846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009039.hg.1 |
NR_106864.1 |
Refseq |