Assay Details
| Gene Symbol: MIR6877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106937.1 | 1 | 32 | 53 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105376306 | LOC105376306 | -21.5kb | Anti-sense | Chr. 9 - 133028152 - 133030422 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100996574 | LOC100996574 | 6.0kb | Anti-sense | Chr. 9 - 133060384 - 133061567 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0900009053.hg.1
Set Membership: |
|

MIR6877
Genomic Map