| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009373.hg.1 |
ENST00000408805.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011268.hg.1 |
ENST00000602324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011268.hg.1 |
ENST00000602324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011268.hg.1 |
ENST00000602324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011268.hg.1 |
NR_108100.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011268.hg.1 |
NR_108100.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011268.hg.1 |
NR_108100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194880.hg.1 |
ENST00000602324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194880.hg.1 |
NR_108100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194880.hg.1 |
TC1900011268.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194880.hg.1 |
ENST00000602324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194880.hg.1 |
NR_108100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011268.hg.1 |
ENST00000602324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011268.hg.1 |
ENST00000602324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011268.hg.1 |
ENST00000602324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011268.hg.1 |
NR_108100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011268.hg.1 |
NR_108100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011268.hg.1 |
NR_108100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194879.hg.1 |
ENST00000602324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194879.hg.1 |
NR_108100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194879.hg.1 |
TC1900011268.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194879.hg.1 |
ENST00000602324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194879.hg.1 |
NR_108100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009373.hg.1 |
ENST00000408805.1 |
Ensembl |