| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008206.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157004.hg.1 |
TC0700008206.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
ENST00000454066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
NR_110077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157003.hg.1 |
TC0700008206.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700157005.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157005.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157005.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157005.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157005.hg.1 |
ENST00000439234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700157005.hg.1 |
NR_110076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700157005.hg.1 |
TC0700008206.hg.1 |
Clariom D TC |