| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010394.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010394.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010394.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010394.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010394.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010394.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010394.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010394.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010394.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010394.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176666.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176666.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176666.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176666.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176666.hg.1 |
TC0100010394.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100176667.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176667.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176667.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176667.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176667.hg.1 |
TC0100010394.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100176670.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176670.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176670.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176670.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176670.hg.1 |
TC0100010394.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010394.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010394.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010394.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010394.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010394.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010394.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010394.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010394.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010394.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010394.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176669.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176669.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176669.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176669.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176669.hg.1 |
TC0100010394.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100176665.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176665.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176665.hg.1 |
ENST00000415437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100176665.hg.1 |
NR_110699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176665.hg.1 |
TC0100010394.hg.1 |
Clariom D TC |