| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
ENST00000587575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
ENST00000587575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
ENST00000587575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
ENST00000592569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
ENST00000592569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
ENST00000592569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
ENST00000592569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
ENST00000592569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009048.hg.1 |
NR_110847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168387.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168387.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168387.hg.1 |
TC1700009048.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700168387.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168387.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168378.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168378.hg.1 |
TC1700009048.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700168378.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168380.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168380.hg.1 |
TC1700009048.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700168380.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168386.hg.1 |
ENST00000587575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168386.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168386.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168386.hg.1 |
TC1700009048.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700168386.hg.1 |
ENST00000587575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168386.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168386.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
ENST00000587575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
ENST00000587575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
ENST00000587575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
ENST00000592569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
ENST00000592569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
ENST00000592569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
ENST00000592569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
ENST00000592569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009048.hg.1 |
NR_110847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168385.hg.1 |
ENST00000587575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168385.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168385.hg.1 |
NR_110846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168385.hg.1 |
TC1700009048.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700168385.hg.1 |
ENST00000587575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168385.hg.1 |
NR_110845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168385.hg.1 |
NR_110846.1 |
Refseq |