Assay Details
| Gene Symbol: LOC101928710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110849.1 | 4 - 5 | 710 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dynein axonemal heavy chain 17 | DNAH17 |
DNAHL1, DNEL2 |
-40.0kb | Sense | Chr. 17 - 78423697 - 78577430 on NCBI Build GRCh38 |
| Downstream | cytohesin 1 | CYTH1 |
B2-1, CYTOHESIN-1, D17S811E, PSCD1, SEC7 |
49.7kb | Sense | Chr. 17 - 78674047 - 78782342 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | TC1700011907.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197187.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | TC1700011907.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197186.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197176.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197176.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197176.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197176.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197176.hg.1 | TC1700011907.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197176.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197176.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197176.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197176.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | TC1700011907.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197188.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011907.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | TC1700011907.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | ENST00000586185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197185.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | TC1700011907.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | ENST00000590023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | NR_110848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197177.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197189.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197189.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197189.hg.1 | NR_110849.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197189.hg.1 | TC1700011907.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197189.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197189.hg.1 | ENST00000591384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197189.hg.1 | NR_110849.1 | Refseq |

LOC101928710
Genomic Map