| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
NR_110879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
NR_110879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
NR_110879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008037.hg.1 |
NR_110879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000509322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
NR_110879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
NR_110879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
NR_110879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008037.hg.1 |
NR_110879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155296.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155296.hg.1 |
NR_110879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155296.hg.1 |
ENST00000511917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155296.hg.1 |
NR_110879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155296.hg.1 |
TC0400008037.hg.1 |
Clariom D TC |