| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008425.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008425.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008425.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008425.hg.1 |
ENST00000617252.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008425.hg.1 |
ENST00000617252.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008425.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008425.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008425.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160138.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160138.hg.1 |
ENST00000617252.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160138.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160138.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160138.hg.1 |
ENST00000617252.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160138.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160138.hg.1 |
TC0300008425.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300160140.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160140.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160140.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160140.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160140.hg.1 |
TC0300008425.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300008425.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008425.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008425.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008425.hg.1 |
ENST00000617252.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008425.hg.1 |
ENST00000617252.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008425.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008425.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008425.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160139.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160139.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160139.hg.1 |
ENST00000487512.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160139.hg.1 |
NR_121607.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160139.hg.1 |
TC0300008425.hg.1 |
Clariom D TC |