| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008217.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008217.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008217.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008217.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008217.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008217.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008217.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008217.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008217.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157478.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157478.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157478.hg.1 |
TC0600008217.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600157487.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600157487.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157487.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600157487.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157487.hg.1 |
TC0600008217.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600157479.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600157479.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157479.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600157479.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157479.hg.1 |
TC0600008217.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600157488.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157488.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157488.hg.1 |
TC0600008217.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600157484.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600157484.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157484.hg.1 |
ENST00000437859.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600157484.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157484.hg.1 |
TC0600008217.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600157475.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157475.hg.1 |
NR_125839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600157475.hg.1 |
TC0600008217.hg.1 |
Clariom D TC |