| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007882.hg.1 |
ENST00000444686.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007882.hg.1 |
ENST00000444686.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007882.hg.1 |
ENST00000444686.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007882.hg.1 |
ENST00000451630.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007882.hg.1 |
ENST00000451630.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007882.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007882.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007882.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300153021.hg.1 |
ENST00000444686.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300153021.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300153021.hg.1 |
ENST00000444686.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300153021.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300153021.hg.1 |
TC1300007882.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300007882.hg.1 |
ENST00000444686.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007882.hg.1 |
ENST00000444686.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007882.hg.1 |
ENST00000444686.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007882.hg.1 |
ENST00000451630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007882.hg.1 |
ENST00000451630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007882.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007882.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007882.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300153024.hg.1 |
ENST00000451630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300153024.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300153024.hg.1 |
ENST00000451630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300153024.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300153024.hg.1 |
TC1300007882.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300153022.hg.1 |
ENST00000444686.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300153022.hg.1 |
ENST00000451630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300153022.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300153022.hg.1 |
ENST00000444686.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300153022.hg.1 |
ENST00000451630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300153022.hg.1 |
NR_125912.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300153022.hg.1 |
TC1300007882.hg.1 |
Clariom D TC |