| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012894.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012894.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012894.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012894.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012894.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012894.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195288.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195288.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195288.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195288.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195288.hg.1 |
TC0100012894.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100195286.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195286.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195286.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195286.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195286.hg.1 |
TC0100012894.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100195287.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195287.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195287.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195287.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195287.hg.1 |
TC0100012894.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100012894.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012894.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012894.hg.1 |
ENST00000376620.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012894.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012894.hg.1 |
NR_126000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012894.hg.1 |
NR_126000.1 |
Refseq |