| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010725.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178764.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100178764.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178764.hg.1 |
ENST00000451341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100178764.hg.1 |
NR_126001.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178764.hg.1 |
TC0100010725.hg.1 |
Clariom D TC |