Assay Details
| Gene Symbol: KRT73-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126005.1 | 2 - 3 | 2065 | 58 | ||
| GenBank mRNA | AK128146.1 | - | 2 - 3 | 2065 | 58 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | keratin 72 | KRT72 |
K6IRS2, K6irs, KRT6, KRT6IRS2 |
-9.4kb | Anti-sense | Chr. 12 - 52585589 - 52602900 on NCBI Build GRCh38 |
| Downstream | keratin 2 | KRT2 |
CK-2e, K2e, KRT2A, KRT2E, KRTE |
29.3kb | Anti-sense | Chr. 12 - 52644558 - 52652175 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007675.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154766.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154766.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154766.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154766.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154766.hg.1 | TC1200007675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200154759.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154759.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154759.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154759.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154759.hg.1 | TC1200007675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200154765.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154765.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154765.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154765.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154765.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154765.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154765.hg.1 | TC1200007675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000305748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000546934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | ENST00000552855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010764.hg.1 | NM_175068.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154761.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154761.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154761.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154761.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154761.hg.1 | TC1200007675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154764.hg.1 | TC1200007675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154760.hg.1 | TC1200007675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | ENST00000549180.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | ENST00000551089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | ENST00000552364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | NR_126005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154762.hg.1 | TC1200007675.hg.1 | Clariom D TC |

KRT73-AS1
Genomic Map