| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009736.hg.1 |
ENST00000449551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009736.hg.1 |
ENST00000449551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009736.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009736.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009736.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164965.hg.1 |
ENST00000449551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164965.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164965.hg.1 |
ENST00000449551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164965.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164965.hg.1 |
TC1300009736.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300164966.hg.1 |
ENST00000449551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164966.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164966.hg.1 |
ENST00000449551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164966.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164966.hg.1 |
TC1300009736.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300009736.hg.1 |
ENST00000449551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009736.hg.1 |
ENST00000449551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009736.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009736.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009736.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164964.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164964.hg.1 |
NR_046848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164964.hg.1 |
TC1300009736.hg.1 |
Clariom D TC |