Assay Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281789.1 | NP_001268718.1 | 1 - 2 | 141 | 95 | |
| NR_104043.1 | 1 - 2 | 141 | 95 | |||
| GenBank mRNA | AK095420.1 | - | 1 - 2 | 114 | 95 | |
| AK307309.1 | - | 1 - 2 | 83 | 95 | ||
| BC022550.1 | - | 1 - 2 | 92 | 95 | ||
| FJ516381.1 | - | 1 - 2 | 69 | 95 | ||
| HY011229.1 | - | 1 - 2 | 142 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK095420.1, BC022550.1, FJ516381.1, NM_001281789.1 transcripts.
Set Membership: |
|

CNEP1R1
Genomic Map