Assay Details
| Gene Symbol: LOC101448202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103451.2 | 2 | 823 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | COL5A1 antisense RNA 1 | COL5A1-AS1 |
C9orf104, bA54A22.4 |
-166.4kb | Sense | Chr. 9 - 134649385 - 134653680 on NCBI Build GRCh38 |
| Downstream | microRNA 3689c | MIR3689C | 29.1kb | Sense | Chr. 9 - 134849298 - 134849369 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101448202
Genomic Map