Assay Details
| Gene Symbol: TMEM53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300746.1 | NP_001287675.1 | 1 - 2 | 282 | 102 | |
| GenBank mRNA | AK296548.1 | - | 1 - 2 | 120 | 102 | |
| AL832539.1 | - | 1 - 2 | 116 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK296548.1, AL832539.1, NM_001300746.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372244.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372244.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372244.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372244.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000372244.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000420706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000420706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000420706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000420706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000495630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000495630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | ENST00000495630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372235.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372237.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372243.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372244.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372244.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372244.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372244.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000372244.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000420706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000420706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000420706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000420706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000468117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000476724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000495630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000495630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | ENST00000495630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300747.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_001300748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013983.hg.1 | NM_024587.3 | Refseq |

TMEM53
Genomic Map