Assay Details
| Gene Symbol: GRAMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146319.1 | NP_001139791.1 | 9 - 10 | 1134 | 82 | |
| NM_001146320.1 | NP_001139792.1 | 8 - 9 | 1171 | 82 | ||
| NM_001146321.1 | NP_001139793.1 | 10 - 11 | 1326 | 82 | ||
| NM_001146322.1 | NP_001139794.1 | 9 - 10 | 945 | 82 | ||
| NM_023927.2 | NP_076416.2 | 9 - 10 | 1283 | 82 | ||
| XM_005272058.3 | XP_005272115.1 | 9 - 10 | 1021 | 82 | ||
| XM_005272063.3 | XP_005272120.1 | 9 - 10 | 1218 | 82 | ||
| XM_011543592.2 | XP_011541894.1 | 10 - 11 | 1995 | 82 | ||
| XM_017009782.1 | XP_016865271.1 | 10 - 11 | 1128 | 82 | ||
| XM_017009783.1 | XP_016865272.1 | 8 - 9 | 739 | 82 | ||
| XM_017009785.1 | XP_016865274.1 | 8 - 9 | 980 | 82 | ||
| GenBank mRNA | AK024966.1 | - | 9 - 10 | 1051 | 82 | |
| AK225829.1 | - | 9 - 10 | 1051 | 82 | ||
| AK293307.1 | - | 8 - 9 | 876 | 82 | ||
| AK296267.1 | - | 10 - 11 | 1031 | 82 | ||
| AK298009.1 | - | 9 - 10 | 868 | 82 | ||
| AK300154.1 | - | 9 - 10 | 1079 | 82 | ||
| AK303857.1 | - | 9 - 10 | 885 | 82 | ||
| AK316288.1 | - | 9 - 10 | 1098 | 82 | ||
| AY116970.1 | - | 9 - 10 | 853 | 82 | ||
| BC008590.1 | - | 9 - 10 | 1016 | 4148772 | 82 | |
More Information
Set Membership: |
|

GRAMD3
Genomic Map