Assay Details
| Gene Symbol: GRAMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146320.1 | NP_001139792.1 | 2 - 3 | 631 | 84 | |
| XM_011543595.2 | XP_011541897.1 | 2 - 3 | 335 | 84 | ||
| XM_017009783.1 | XP_016865272.1 | 2 - 3 | 199 | 84 | ||
| XM_017009784.1 | XP_016865273.1 | 2 - 3 | 387 | 84 | ||
| XM_017009785.1 | XP_016865274.1 | 2 - 3 | 440 | 84 | ||
| GenBank mRNA | AK293307.1 | - | 2 - 3 | 336 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK293307.1, NM_001146320.1, XM_011543595.2, XM_017009783.1, XM_017009784.1, XM_017009785.1 transcripts.
Set Membership: |
|

GRAMD3
Genomic Map