Assay Details
| Gene Symbol: EGR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199880.1 | NP_001186809.1 | 1 - 2 | 88 | 111 | |
| XM_005273426.3 | XP_005273483.1 | 2 - 3 | 136 | 111 | ||
| XM_011544429.2 | XP_011542731.1 | 2 - 3 | 248 | 111 | ||
| GenBank mRNA | AK295134.1 | - | 1 - 2 | 89 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK295134.1, NM_001199880.1, XM_005273426.3 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of endothelial cell proliferation
cell migration involved in sprouting angiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
neuromuscular synaptic transmission
peripheral nervous system development
muscle organ development
circadian rhythm
positive regulation of T cell differentiation in thymus
endothelial cell chemotaxis
cellular response to vascular endothelial growth factor stimulus
negative regulation of apoptotic process
cellular response to fibroblast growth factor stimulus
regulation of gamma-delta T cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
positive regulation of endothelial cell proliferation
cell migration involved in sprouting angiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
neuromuscular synaptic transmission
peripheral nervous system development
muscle organ development
circadian rhythm
positive regulation of T cell differentiation in thymus
endothelial cell chemotaxis
cellular response to vascular endothelial growth factor stimulus
negative regulation of apoptotic process
cellular response to fibroblast growth factor stimulus
regulation of gamma-delta T cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000519624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000519624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000519624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000521141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000521141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000521141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000521141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000523627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000523627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000523627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000523627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006994.hg.1 | ENST00000523627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000518773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000518773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000524088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000524088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000524088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | ENST00000524088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000317216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000518773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000518773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000519492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000522910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000524088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000524088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000524088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | ENST00000524088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_001199881.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009856.hg.1 | NM_004430.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000519624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000519624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000519624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000521141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000521141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000521141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000521141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000523627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000523627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000523627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000523627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006994.hg.1 | ENST00000523627.1 | Ensembl |

EGR3
Genomic Map