| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000469440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000469440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000469440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000469440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000469440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000496807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000496807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000496807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
ENST00000496807.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000343837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000469440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000469440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000469440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000469440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000469440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000496807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000496807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000496807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
ENST00000496807.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007673.hg.1 |
NM_001080537.1 |
Refseq |