Assay Details
| Gene Symbol: BAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172415.1 | NP_001165886.1 | 3 - 4 | 752 | 86 | |
| NM_004323.5 | NP_004314.5 | 3 - 4 | 752 | 86 | ||
| GenBank mRNA | AF022224.1 | - | 3 - 4 | 710 | 86 | |
| AF116273.1 | - | 3 - 4 | 680 | 86 | ||
| AK096038.1 | - | 4 - 5 | 515 | 86 | ||
| AK222749.1 | - | 3 - 4 | 699 | 86 | ||
| AK316577.1 | - | 3 - 4 | 678 | 86 | ||
| BC001936.1 | - | 3 - 4 | 713 | 2823774 | 86 | |
| BC014774.1 | - | 3 - 4 | 562 | 86 | ||
| KU177932.1 | - | 3 - 4 | 452 | 86 | ||
| KU177933.1 | - | 4 - 5 | 594 | 86 | ||
| U46917.1 | - | 3 - 4 | 712 | 86 | ||
| Z35491.1 | - | 3 - 4 | 730 | 86 | ||
More Information
Set Membership: |
|

BAG1
Genomic Map