Assay Details
| Gene Symbol: BAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172415.1 | NP_001165886.1 | 6 - 7 | 1038 | 101 | |
| NM_004323.5 | NP_004314.5 | 6 - 7 | 1038 | 101 | ||
| GenBank mRNA | AF022224.1 | - | 6 - 7 | 996 | 101 | |
| AK093238.1 | - | 3 - 4 | 1697 | 101 | ||
| AK096038.1 | - | 7 - 8 | 801 | 101 | ||
| AK222749.1 | - | 6 - 7 | 985 | 101 | ||
| AK316577.1 | - | 6 - 7 | 964 | 101 | ||
| BC001936.1 | - | 6 - 7 | 999 | 2823774 | 101 | |
| BC014774.1 | - | 6 - 7 | 848 | 101 | ||
| KU177932.1 | - | 6 - 7 | 738 | 101 | ||
| KU177933.1 | - | 7 - 8 | 880 | 101 | ||
| U46917.1 | - | 6 - 7 | 998 | 101 | ||
| Z35491.1 | - | 6 - 7 | 1016 | 101 | ||
More Information
Set Membership: |
|

BAG1
Genomic Map