Assay Details
| Gene Symbol: LOC101928098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132786.1 | 1 - 2 | 595 | 104 | ||
| GenBank mRNA | BC040907.1 | - | 1 - 2 | 595 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hCG2028352-like | LOC729970 | -98.3kb | Sense | Chr. 1 - 94928028 - 94963270 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 56 | TMEM56 | 25.0kb | Sense | Chr. 1 - 95092517 - 95197607 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928098
Genomic Map