Assay Details
| Gene Symbol: KLK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015596.1 | NP_056411.1 | 1 - 2 | 95 | 73 | |
| GenBank mRNA | AB108824.1 | - | 1 - 2 | 52 | 73 | |
| AB451465.1 | - | 1 - 2 | 52 | 73 | ||
| AL050220.1 | - | 1 - 2 | 100 | 73 | ||
| AY923171.1 | - | 1 - 2 | 95 | 73 | ||
| BC069334.1 | - | 1 - 2 | 96 | 7262205 | 73 | |
| BC069543.1 | - | 1 - 2 | 95 | 7262217 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kallikrein related peptidase 12 | KLK12 |
KLK-L5, KLKL5 |
-24.8kb | Sense | Chr. 19 - 51028954 - 51035651 on NCBI Build GRCh38 |
| Downstream | kallikrein related peptidase 14 | KLK14 | KLK-L6 | 12.4kb | Sense | Chr. 19 - 51077495 - 51084210 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

KLK13
Genomic Map