| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010695.hg.1 |
ENST00000561964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010695.hg.1 |
ENST00000561964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010695.hg.1 |
ENST00000561964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010695.hg.1 |
ENST00000561964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010695.hg.1 |
ENST00000566676.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010695.hg.1 |
ENST00000566676.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010695.hg.1 |
ENST00000566676.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010695.hg.1 |
ENST00000566676.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010695.hg.1 |
ENST00000566676.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006560.hg.1 |
ENST00000566797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006560.hg.1 |
ENST00000566797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006560.hg.1 |
ENST00000566797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006560.hg.1 |
ENST00000566797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006560.hg.1 |
ENST00000566797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006560.hg.1 |
NR_130780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006560.hg.1 |
NR_130780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006560.hg.1 |
NR_130780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006560.hg.1 |
NR_130780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006560.hg.1 |
NR_130780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010695.hg.1 |
ENST00000561964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010695.hg.1 |
ENST00000561964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010695.hg.1 |
ENST00000561964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010695.hg.1 |
ENST00000561964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010695.hg.1 |
ENST00000566676.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010695.hg.1 |
ENST00000566676.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010695.hg.1 |
ENST00000566676.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010695.hg.1 |
ENST00000566676.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010695.hg.1 |
ENST00000566676.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006560.hg.1 |
ENST00000566797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006560.hg.1 |
ENST00000566797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006560.hg.1 |
ENST00000566797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006560.hg.1 |
ENST00000566797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006560.hg.1 |
ENST00000566797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006560.hg.1 |
NR_130780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006560.hg.1 |
NR_130780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006560.hg.1 |
NR_130780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006560.hg.1 |
NR_130780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006560.hg.1 |
NR_130780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500010696.hg.1 |
ENST00000565893.1 |
Ensembl |