Assay Details
| Gene Symbol: USP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020718.3 | NP_065769.3 | 16 | 10123 | 81 | |
| XM_017023496.1 | XP_016878985.1 | 15 | 10034 | 81 | ||
| XM_017023497.1 | XP_016878986.1 | 16 | 11649 | 81 | ||
| XM_017023498.1 | XP_016878987.1 | 15 | 9436 | 81 | ||
| XM_017023499.1 | XP_016878988.1 | 13 | 9240 | 81 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007214.hg.1 | ENST00000624328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007214.hg.1 | ENST00000624328.1 | Ensembl |

USP31
Genomic Map