Assay Details
| Gene Symbol: TMEM99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005257089.4 | XP_005257146.1 | 1 - 2 | 47 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005257089.4 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000496847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_145274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_145274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_145274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_145274.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_145274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000496847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_145274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_145274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_145274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_145274.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_145274.3 | Refseq |

TMEM99
Genomic Map