Assay Details
| Gene Symbol: SPRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128210.1 | NP_001121682.1 | 3 - 4 | 556 | 95 | |
| NM_181784.2 | NP_861449.2 | 3 - 4 | 909 | 95 | ||
| XM_005264200.4 | XP_005264257.2 | 3 - 4 | 986 | 95 | ||
| XM_005264202.4 | XP_005264259.1 | 3 - 4 | 986 | 95 | ||
| GenBank mRNA | AK056479.1 | - | 3 - 4 | 473 | 95 | |
| AK299079.1 | - | 3 - 4 | 409 | 95 | ||
| AY299090.1 | - | 3 - 4 | 570 | 95 | ||
| BC029300.1 | - | 3 - 4 | 486 | 95 | ||
| BC111495.1 | - | 3 - 4 | 897 | 95 | ||
| BC130292.1 | - | 3 - 4 | 506 | 40146323 | 95 | |
| BC136334.1 | - | 3 - 4 | 506 | 9020321 | 95 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
multicellular organism development
fibroblast growth factor receptor signaling pathway
negative regulation of peptidyl-threonine phosphorylation
positive regulation of DNA damage response, signal transduction by p53 class mediator
regulation of protein deacetylation
stem cell factor receptor binding
protein binding
protein kinase binding
protein serine/threonine kinase inhibitor activity
inactivation of MAPK activity
multicellular organism development
fibroblast growth factor receptor signaling pathway
negative regulation of peptidyl-threonine phosphorylation
positive regulation of DNA damage response, signal transduction by p53 class mediator
regulation of protein deacetylation
stem cell factor receptor binding
protein binding
protein kinase binding
protein serine/threonine kinase inhibitor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000421087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000421087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000421087.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000426832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000426832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000426832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000426832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000427238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000427238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000427238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000356388.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000421087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000421087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000421087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000426832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000426832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000426832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000426832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000427238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000427238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000427238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000440972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000443619.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000452315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | ENST00000474228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_001128210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012863.hg.1 | NM_181784.2 | Refseq |

SPRED2
Genomic Map