| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000427417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000427417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000427417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000427417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000427417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000442699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000442699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000442699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000444166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000444166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000444166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000444166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000444166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000445644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000445644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000445644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000445644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000445644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000455659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000455659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000455659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000455659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000458337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000458337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000458337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000458337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000458337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000311832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000427417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000427417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000427417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000427417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000427417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000442699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000442699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000442699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000444166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000444166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000444166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000444166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000444166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000445644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000445644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000445644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000445644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000445644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000455659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000455659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000455659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000455659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000458337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000458337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000458337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000458337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000458337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000471704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
ENST00000472729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007739.hg.1 |
NM_152516.3 |
Refseq |