Assay Details
Gene Symbol: PDZRN3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046681.1 | 1 - 2 | 401 | 90 | ||
GenBank mRNA | AL692084.1 | - | 1 - 2 | 402 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000619517.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000619517.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000619517.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000619517.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000478988.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608304.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000608743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000619517.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000619517.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000619517.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000619517.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | ENST00000625169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007849.hg.1 | NR_046681.1 | Refseq |
