Assay Details
| Gene Symbol: KLHL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC015962.1 | - | 1 - 2 | 138 | 67 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |

KLHL18
Genomic Map