Assay Details
| Gene Symbol: WNT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017007127.1 | XP_016862616.1 | 2 - 3 | 249 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017007127.1 transcripts.
Set Membership: |
|

WNT5A
Genomic Map