Assay Details
| Gene Symbol: WNT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534085.2 | XP_011532387.1 | 1 - 2 | 1835 | 85 | |
| XM_011534086.2 | XP_011532388.1 | 1 - 2 | 1835 | 85 | ||
| XM_017007127.1 | XP_016862616.1 | 3 - 4 | 338 | 85 | ||
| XM_017007128.1 | XP_016862617.1 | 1 - 2 | 1835 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011534085.2, XM_011534086.2, XM_017007127.1, XM_017007128.1 transcripts.
Set Membership: |
|

WNT5A
Genomic Map