| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000473093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000473093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000473093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000481989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000481989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000481989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000481989.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000491050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000491050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000491050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
ENST00000491050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000309993.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000434835.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000473093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000473093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000473093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000481989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000481989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000481989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000481989.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000491050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000491050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000491050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
ENST00000491050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_001145417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012527.hg.1 |
NM_018133.3 |
Refseq |