Assay Details
| Gene Symbol: TMEM45B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017017189.1 | XP_016872678.1 | 1 - 2 | 94 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017017189.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000525709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000525709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000525709.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000529381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000529381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | ENST00000529381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006985.hg.1 | ENST00000558967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006985.hg.1 | ENST00000558967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006985.hg.1 | ENST00000558967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006985.hg.1 | ENST00000558967.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006985.hg.1 | ENST00000560178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006985.hg.1 | ENST00000560178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006985.hg.1 | ENST00000560178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000281441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000524567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000525709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000525709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000525709.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000527754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000529381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000529381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | ENST00000529381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009514.hg.1 | NM_138788.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006985.hg.1 | ENST00000558967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006985.hg.1 | ENST00000558967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006985.hg.1 | ENST00000558967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006985.hg.1 | ENST00000558967.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006985.hg.1 | ENST00000560178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006985.hg.1 | ENST00000560178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006985.hg.1 | ENST00000560178.1 | Ensembl |

TMEM45B
Genomic Map