Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |