Assay Details
| Gene Symbol: LOC101928521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC020343.2 | - | 1 - 2 | 566 | 59 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000489312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000489312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000489312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000489312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000489312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000598370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000598370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000598370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000598370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000598370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000468356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000489312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000489312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000489312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000489312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000489312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000598370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000598370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000598370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000598370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000598370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009692.hg.1 | ENST00000599030.1 | Ensembl |

LOC101928521
Genomic Map