Assay Details
Gene Symbol: SGCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000337.5 | NP_000328.2 | 9 | 3604 | 111 | |
NM_001128209.1 | NP_001121681.1 | 8 | 3558 | 111 | ||
GenBank mRNA | AK129889.1 | - | 1 | 1123 | 111 | |
BX537948.1 | - | 8 | 3569 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000337851.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000435422.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000517913.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | ENST00000524347.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_000337.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_001128209.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009192.hg.1 | NM_172244.2 | Refseq |
