Assay Details
| Gene Symbol: SIAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199922.1 | NP_001186851.1 | 3 - 4 | 375 | 89 | |
| GenBank mRNA | AF300796.1 | - | 3 - 4 | 375 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF300796.1, NM_001199922.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |

SIAE
Genomic Map