Assay Details
| Gene Symbol: PKD2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011540323.2 | XP_011538625.1 | 1 - 2 | 124 | 67 | |
| XM_011540325.2 | XP_011538627.1 | 1 - 2 | 124 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011540323.2, XM_011540325.2 transcripts.
Set Membership: |
|

PKD2L1
Genomic Map