Assay Details
| Gene Symbol: PKD2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011540323.2 | XP_011538625.1 | 2 - 3 | 244 | 68 | |
| XM_011540325.2 | XP_011538627.1 | 2 - 3 | 244 | 68 | ||
| XM_017016875.1 | XP_016872364.1 | 2 - 3 | 192 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011540323.2, XM_011540325.2, XM_017016875.1 transcripts.
Set Membership: |
|

PKD2L1
Genomic Map