Assay Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK311157.1 | - | 2 - 3 | 187 | 141 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000319285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000565758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000567983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | ENST00000568973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | NM_024096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009966.hg.1 | NM_024096.1 | Refseq |

ZNF771
Genomic Map