Assay Details
| Gene Symbol: SH3TC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024577.3 | NP_078853.2 | 17 | 26489 | 65 | |
| GenBank mRNA | AF086088.1 | - | 1 | 218 | 65 | |
| AK023641.1 | - | 1 | 1875 | 65 | ||
More Information
Set Membership: |
|

SH3TC2
Genomic Map